Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP63 SIRNA Assays ›
Gene Symbol:
CEP63
Gene Name:
centrosomal protein 63
Gene Aliases:
SCKL6
Chromosome Location:
Chr.3: 134485733 - 134629607 on Build GRCh38
UniGene ID:
Hs.443301
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
32501
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP63
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042383.1
NP_001035842.1
3
486
NM_001042384.1
NP_001035843.1
3
486
NM_001042400.1
NP_001035859.1
3
314
NM_025180.3
NP_079456.2
4
590
XM_005247795.4
XP_005247852.1
3
263
XM_005247797.3
XP_005247854.1
3
518
XM_005247799.3
XP_005247856.1
3
313
XM_006713760.3
XP_006713823.1
3
195
XM_011513194.2
XP_011511496.1
3
312
XM_017007244.1
XP_016862733.1
3
306
XM_017007245.1
XP_016862734.1
3
303
XM_017007246.1
XP_016862735.1
3
195
XM_017007247.1
XP_016862736.1
3
312
XM_017007248.1
XP_016862737.1
3
479
XM_017007249.1
XP_016862738.1
4
568
XM_017007250.1
XP_016862739.1
3
304
XM_017007251.1
XP_016862740.1
3
302
XM_017007252.1
XP_016862741.1
3
219
XM_017007253.1
XP_016862742.1
3
298
XM_017007254.1
XP_016862743.1
3
195
XM_017007255.1
XP_016862744.1
3
518
XM_017007256.1
XP_016862745.1
4
568
XM_017007257.1
XP_016862746.1
3
300
XM_017007258.1
XP_016862747.1
3
195
XM_017007259.1
XP_016862748.1
3
312
XM_017007260.1
XP_016862749.1
3
204
XM_017007261.1
XP_016862750.1
3
263
XM_017007262.1
XP_016862751.1
3
312
XM_017007263.1
XP_016862752.1
3
518
XM_017007264.1
XP_016862753.1
4
568
XM_017007265.1
XP_016862754.1
3
296
XM_017007266.1
XP_016862755.1
3
312
XM_017007267.1
XP_016862756.1
3
479
XM_017007268.1
XP_016862757.1
3
483
XM_017007269.1
XP_016862758.1
4
568
GenBank mRNA
AK023448.1
-
3
260
AK023738.1
-
3
427
AK056465.1
-
4
568
BC014050.2
-
3
475
3639744
BC068997.1
-
3
234
5951988
BI561425.1
-
3
317
CX751767.1
-
3
426
DB011963.1
-
3
519
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage checkpoint
G2/M transition of mitotic cell cycle
mitotic nuclear division
centriole replication
signal transduction in response to DNA damage
spindle assembly
cell division
de novo centriole assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-858577f6d7-cpd7z:80/100.66.134.75:80.
git-commit: 8a4c7f5d60a2609f21a9257632c46d3db2b553cc
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0