Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199573.1 | NP_001186502.1 | 2 | 519 | ||
NM_006074.4 | NP_006065.2 | 2 | 519 | |||
GenBank mRNA | AK290434.1 | - | 2 | 249 | ||
AK298715.1 | - | 2 | 305 | |||
AK301192.1 | - | 2 | 224 | |||
AK308221.1 | - | 2 | 339 | |||
AK308796.1 | - | 2 | 235 | |||
AK310098.1 | - | 2 | 339 | |||
BC022281.1 | - | 2 | 326 | 4718024 | ||
BC035582.1 | - | 2 | 292 | 5583800 | ||
CD621768.1 | - | 2 | 420 | |||
DC364428.1 | - | 2 | 496 | |||
HQ842635.1 | - | 1 | 140 | |||
X82200.1 | - | 2 | 262 |