Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GAL3ST1 SIRNA Assays ›
          Gene Symbol:
          GAL3ST1
          Gene Name:
          galactose-3-O-sulfotransferase 1
          Gene Aliases:
          CST
          Chromosome Location:
          Chr.22: 30554635 - 30574588 on Build GRCh38
          UniGene ID:
          Hs.17958
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 289496

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GAL3ST1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001318103.1 NP_001305032.1 3 607
          NM_001318104.1 NP_001305033.1 4 717
          NM_001318105.1 NP_001305034.1 3 668
          NM_001318106.1 NP_001305035.1 4 793
          NM_001318107.1 NP_001305036.1 4 781
          NM_001318108.1 NP_001305037.1 4 778
          NM_001318109.1 NP_001305038.1 3 552
          NM_001318110.1 NP_001305039.1 4 685
          NM_001318111.1 NP_001305040.1 3 575
          NM_001318112.1 NP_001305041.1 3 521
          NM_001318113.1 NP_001305042.1 4 631
          NM_001318114.1 NP_001305043.1 3 524
          NM_001318115.1 NP_001305044.1 4 650
          NM_001318116.1 NP_001305045.1 5 773
          NM_004861.2 NP_004852.1 4 704
          XM_011530518.2 XP_011528820.1 3 2142
          XM_011530522.2 XP_011528824.1 3 2258
          XM_011530524.1 XP_011528826.1 3 611
          XM_011530528.1 XP_011528830.1 3 512
          XM_017029096.1 XP_016884585.1 3 728
          XM_017029097.1 XP_016884586.1 3 2026
          XM_017029098.1 XP_016884587.1 4 2151
          GenBank mRNA AK313097.1 - 3 629
          BC014649.1 - 4 611 4564703
          BC019077.2 - 3 506 4562507
          BM923857.1 - 4 717
          BP220526.1 - 3 547
          BP276494.1 - 3 521
          CR456350.1 - 3 549
          D88667.1 - 4 679
          DA371478.1 - 3 562
          DA372454.1 - 3 598
          DA381099.1 - 3 521

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein N-linked glycosylation
          galactosylceramide biosynthetic process
          spermatogenesis
          myelination
          galactosylceramide sulfotransferase activity
          sulfotransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7f76987d59-9ndr4:80/100.66.134.75:80.
          git-commit: fa11bf362e7d47d57dd0debebfa11e830f891645
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0