Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MIER1 SIRNA Assays ›
          Gene Symbol:
          MIER1
          Gene Name:
          MIER1 transcriptional regulator
          Gene Aliases:
          ER1, MI-ER1
          Chromosome Location:
          Chr.1: 66924895 - 66988619 on Build GRCh38
          UniGene ID:
          Hs.605432
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 28816

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MIER1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077700.2 NP_001071168.2 3, 4 335
          NM_001077701.2 NP_001071169.1 2, 3 191
          NM_001077702.2 NP_001071170.2 4, 5 409
          NM_001077703.2 NP_001071171.2 3, 4 335
          NM_001077704.2 NP_001071172.1 2, 3 191
          NM_001146110.1 NP_001139582.1 4, 5 412
          NM_001146111.1 NP_001139583.1 4, 5 409
          NM_001146112.1 NP_001139584.1 4, 5 370
          NM_001278215.1 NP_001265144.1 4, 5 409
          NM_020948.3 NP_065999.2 4, 5 409
          XM_005271076.3 XP_005271133.1 4, 5 447
          XM_011541864.2 XP_011540166.1 5, 6 354
          XM_011541865.2 XP_011540167.1 5, 6 315
          XM_017001919.1 XP_016857408.1 5, 6 331
          XM_017001920.1 XP_016857409.1 5, 6 348
          XM_017001921.1 XP_016857410.1 5, 6 376
          XM_017001922.1 XP_016857411.1 5, 6 351
          XM_017001924.1 XP_016857413.1 4, 5 303
          XM_017001925.1 XP_016857414.1 4, 5 408
          XM_017001926.1 XP_016857415.1 5, 6 347
          XM_017001927.1 XP_016857416.1 3, 4 220
          XM_017001928.1 XP_016857417.1 3, 4 296
          XM_017001929.1 XP_016857418.1 4, 5 277
          XM_017001930.1 XP_016857419.1 3, 4 221
          XM_017001931.1 XP_016857420.1 4, 5 255
          GenBank mRNA AF515446.1 - 2, 3 191
          AF515447.1 - 3, 4 272
          AF515448.2 - 4, 5 346
          AK302061.1 - 3, 4 335
          AK307717.1 - 4, 5 306
          AL831987.2 - 4, 5 351
          AY124186.2 - 4, 5 346
          AY124187.1 - 3, 4 272
          AY124188.1 - 2, 3 191
          AY124189.2 - 4, 5 346
          AY124190.1 - 3, 4 272
          AY124191.1 - 2, 3 191
          AY124192.2 - 4, 5 346
          AY124193.1 - 3, 4 272
          AY124194.1 - 2, 3 191
          BC017423.1 - 2, 3 168
          BC108726.1 - 4, 5 290 5194296
          CR627441.1 - 4, 5 246

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          signal transduction
          positive regulation of chromatin silencing
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          DNA binding
          signal transducer activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0