Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FOXP1 SIRNA Assays ›
          Gene Symbol:
          Foxp1
          Gene Name:
          forkhead box P1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 131362178 - 131745135 on Build Rnor_6.0
          UniGene ID:
          Rn.33321
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 281582

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Foxp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001034131.1 NP_001029303.1 8 1145
          XM_006236954.2 XP_006237016.1 7 871
          XM_006236955.2 XP_006237017.1 7 1386
          XM_006236956.1 XP_006237018.1 7 808
          XM_008763148.1 XP_008761370.1 10 1887
          XM_008763149.1 XP_008761371.1 10 1289
          XM_008763150.1 XP_008761372.1 11 1364
          XM_008763151.1 XP_008761373.1 10 1481
          XM_008763152.1 XP_008761374.1 9 1467
          XM_008763153.1 XP_008761375.1 9 2013
          XM_008763154.1 XP_008761376.1 10 1884
          XM_008763155.1 XP_008761377.1 10 1885
          XM_008763156.1 XP_008761378.1 10 1888
          XM_008763157.1 XP_008761379.1 10 1286
          XM_008763158.1 XP_008761380.1 10 1289
          GenBank mRNA BC100267.1 - 8 1145 7383433

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001197321, NM_001197322, NM_053202, AB221602, AK077062, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          positive regulation of mesenchymal cell proliferation
          pre-B cell differentiation
          positive regulation of immunoglobulin production
          negative regulation of B cell apoptotic process
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          heart development
          skeletal muscle tissue development
          motor neuron axon guidance
          positive regulation of endothelial cell migration
          ventral spinal cord development
          osteoclast differentiation
          lung development
          response to lipopolysaccharide
          regulation of tumor necrosis factor production
          immunoglobulin V(D)J recombination
          chemokine (C-C motif) ligand 2 secretion
          osteoclast development
          macrophage activation
          monocyte activation
          endothelial cell activation
          sarcomere organization
          regulation of monocyte differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of smooth muscle cell proliferation
          smooth muscle tissue development
          positive regulation of epithelial cell proliferation
          regulation of interleukin-1 beta secretion
          regulation of inflammatory response
          cardiac muscle cell differentiation
          regulation of cardiac muscle cell proliferation
          negative regulation of androgen receptor signaling pathway
          lung secretory cell differentiation
          T follicular helper cell differentiation
          cardiovascular system development
          interleukin-21 secretion
          regulation of defense response to bacterium
          regulation of lung goblet cell differentiation
          negative regulation of lung goblet cell differentiation
          regulation of macrophage colony-stimulating factor production
          regulation of endothelial tube morphogenesis
          positive regulation of cardiac muscle cell differentiation
          regulation of interleukin-12 secretion
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein homodimerization activity
          metal ion binding
          protein heterodimerization activity
          androgen receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0