Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF415 SIRNA Assays ›
          Gene Symbol:
          ZNF415
          Gene Name:
          zinc finger protein 415
          Gene Aliases:
          ZfLp
          Chromosome Location:
          Chr.19: 53107879 - 53133077 on Build GRCh38
          UniGene ID:
          Hs.147765
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 27030

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF415
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136038.2 NP_001129510.2 4 2133
          NM_001164309.1 NP_001157781.1 4 2006
          NM_018355.3 NP_060825.2 4 1901
          NR_028343.1 7 2262
          XM_006723267.1 XP_006723330.1 6 2219
          XM_006723268.1 XP_006723331.1 4 1967
          XM_006723272.2 XP_006723335.1 5 1999
          XM_006723273.2 XP_006723336.1 5 2083
          XM_006723274.2 XP_006723337.1 5 2160
          XM_006723277.2 XP_006723340.1 5 2094
          XM_006723279.1 XP_006723342.1 5 2253
          XM_006723280.2 XP_006723343.1 2 1709
          XM_006723281.2 XP_006723344.1 3 1735
          XM_006723282.1 XP_006723345.1 3 2044
          XM_011527100.1 XP_011525402.1 5 2243
          XM_011527101.2 XP_011525403.1 5 2218
          XM_011527103.1 XP_011525405.1 5 2038
          XM_011527104.1 XP_011525406.1 5 2332
          XM_017026952.1 XP_016882441.1 5 2046
          XM_017026953.1 XP_016882442.1 6 2366
          XM_017026954.1 XP_016882443.1 6 2243
          XM_017026955.1 XP_016882444.1 6 2194
          XM_017026956.1 XP_016882445.1 7 2344
          XM_017026957.1 XP_016882446.1 6 2082
          XM_017026958.1 XP_016882447.1 7 2398
          XM_017026959.1 XP_016882448.1 6 2269
          XM_017026960.1 XP_016882449.1 5 1984
          XM_017026961.1 XP_016882450.1 6 2430
          XM_017026962.1 XP_016882451.1 6 2275
          XM_017026963.1 XP_016882452.1 5 2171
          XM_017026964.1 XP_016882453.1 5 2121
          XM_017026965.1 XP_016882454.1 4 2135
          XM_017026966.1 XP_016882455.1 5 2263
          XM_017026967.1 XP_016882456.1 4 2061
          XM_017026968.1 XP_016882457.1 6 2359
          XM_017026969.1 XP_016882458.1 2 1780
          XM_017026970.1 XP_016882459.1 6 2236
          XM_017026971.1 XP_016882460.1 6 2455
          XM_017026972.1 XP_016882461.1 4 2037
          XM_017026973.1 XP_016882462.1 5 2229
          XM_017026974.1 XP_016882463.1 6 2308
          GenBank mRNA AF038179.1 - 1 709
          AK002053.1 - 4 1873
          AK095504.1 - 7 2234
          AY283600.1 - 4 1873
          BC063880.1 - 5 2084 30388597
          BC069668.1 - 5 2083 7262632
          DQ925695.1 - 3 1735
          DQ925696.1 - 5 2072
          DQ925697.1 - 6 2082
          DQ925698.1 - 5 2184

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c48b94556-6l4lq:80/100.66.134.75:80.
          git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0