Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126388.1 | 1 | 39 | |||
NR_126389.1 | 1 | 39 | ||||
GenBank mRNA | CN413844.1 | - | 1 | 12 |
Gene Symbol: ERVH-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU669866.1 | - | 2 | 273 |
Gene Symbol: ESRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027122.1 | 1 | 105 | |||
GenBank mRNA | DQ445779.2 | - | 1 | 105 | ||
KM363598.1 | - | 1 | 105 |
Gene Symbol: LOC100126447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321413.1 | NP_001308342.1 | 1 | 87 | ||
GenBank mRNA | EF565112.2 | - | 1 | 87 |
Gene Symbol: LOC101928254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110182.1 | 1 | 101 | |||
NR_110183.1 | 1 | 101 | ||||
GenBank mRNA | CV808846.1 | - | 1 | 84 | ||
EF565094.1 | - | 1 | 102 |
Gene Symbol: MGC32805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051996.1 | 2 | 155 | |||
GenBank mRNA | BC029465.1 | - | 2 | 155 |