Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RHOT1 SIRNA Assays ›
          Gene Symbol:
          RHOT1
          Gene Name:
          ras homolog family member T1
          Gene Aliases:
          ARHT1, MIRO-1, MIRO1
          Chromosome Location:
          Chr.17: 32142454 - 32253374 on Build GRCh38
          UniGene ID:
          Hs.655325
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 26615

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RHOT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001033566.2 NP_001028738.1 4, 5 469
          NM_001033567.2 NP_001028739.2 5, 6 593
          NM_001033568.2 NP_001028740.1 4, 5 469
          NM_001288754.1 NP_001275683.1 4, 5 469
          NM_001288755.1 NP_001275684.1 4, 5 465
          NM_001288758.1 NP_001275687.1 5, 6 593
          NM_018307.4 NP_060777.3 4, 5 469
          NR_110083.1 4, 5 469
          XM_011524969.2 XP_011523271.1 3, 4 193
          XM_011524971.2 XP_011523273.2 4, 5 265
          XM_011524973.2 XP_011523275.2 4, 5 265
          XM_011524975.2 XP_011523277.1 5, 6 722
          XM_011524976.2 XP_011523278.2 4, 5 265
          XM_017024819.1 XP_016880308.1 5, 6 497
          XM_017024820.1 XP_016880309.1 3, 4 193
          XM_017024821.1 XP_016880310.1 4, 5 265
          XM_017024822.1 XP_016880311.1 4, 5 494
          XM_017024823.1 XP_016880312.1 4, 5 265
          XM_017024824.1 XP_016880313.1 5, 6 600
          XM_017024825.1 XP_016880314.1 5, 6 692
          GenBank mRNA AJ496730.1 - 4, 5 457
          AJ517412.1 - 4, 5 230
          AK294407.1 - 4, 5 243
          AL136929.1 - 4, 5 293
          BC041114.1 - 4, 5 403
          BC051818.1 - 3, 4 180
          BC060781.1 - 4, 5 469 4823115
          BC068463.1 - 4, 5 357
          BC125104.1 - 4, 5 298 40118340
          BC125105.1 - 4, 5 298 40118341
          CN352790.1 - 4, 5 426

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          cellular homeostasis
          mitochondrion transport along microtubule
          regulation of small GTPase mediated signal transduction
          mitochondrial outer membrane permeabilization
          molecular_function
          GTPase activity
          calcium ion binding
          protein binding
          GTP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0