Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034054.1 | 4 | 621 | |||
GenBank mRNA | AK096368.1 | - | 4 | 621 | ||
AK311420.1 | - | 3 | 398 | |||
BC035722.1 | - | 4 | 446 |