Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNMAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103149.1 | NP_001096619.1 | 3 | 1452 | ||
NM_018215.3 | NP_060685.2 | 3 | 1657 | |||
XM_011527067.2 | XP_011525369.1 | 3 | 1742 | |||
GenBank mRNA | AK001643.1 | - | 2 | 676 | ||
AK096326.1 | - | 3 | 1217 | |||
AK290218.1 | - | 4 | 1540 | |||
AL834344.1 | - | 3 | 1296 | |||
BC026026.1 | - | 2 | 690 | |||
BC032508.1 | - | 3 | 1635 | 5246377 | ||
BC051688.1 | - | 3 | 1614 | 5295669 | ||
BG708870.1 | - | 2 | 135 | |||
BX649091.1 | - | 2 | 494 |