Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caprin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012185.2 | NP_001012185.2 | 19 | 2201 | ||
XM_006234652.2 | XP_006234714.1 | 20 | 2256 | |||
GenBank mRNA | AY155572.1 | - | 18 | 2057 | ||
BC087123.1 | - | 18 | 2071 |