Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2Y1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001657.1 | NP_001001657.1 | 1 | 521 | ||
GenBank mRNA | BC140756.1 | - | 1 | 562 | 9021624 |