Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000982.1 | NP_001000982.1 | 1 | 787 |
Gene Symbol: LOC103690299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766036.1 | XP_008764258.1 | 1 | 790 |
Gene Symbol: LOC103693025 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766035.1 | XP_008764257.1 | 1 | 745 |
Gene Symbol: LOC103693026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766037.1 | XP_008764259.1 | 2 | 967 |
Gene Symbol: LOC690821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766034.1 | XP_008764256.1 | 1 | 745 |