Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VEZT SIRNA Assays ›
          Gene Symbol:
          VEZT
          Gene Name:
          vezatin, adherens junctions transmembrane protein
          Gene Aliases:
          VEZATIN
          Chromosome Location:
          Chr.12: 95217746 - 95302799 on Build GRCh38
          UniGene ID:
          Hs.24135
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 25121

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: VEZT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017599.3 NP_060069.3 12 2660
          NR_038241.1 13 2733
          NR_038242.1 12 2668
          XM_006719477.3 XP_006719540.1 12 2672
          XM_006719478.2 XP_006719541.1 11 2566
          XM_006719479.2 XP_006719542.1 13 2743
          XM_006719480.3 XP_006719543.1 13 2720
          XM_006719481.2 XP_006719544.1 12 2590
          XM_006719482.3 XP_006719545.1 12 2971
          XM_006719483.3 XP_006719546.1 12 2988
          XM_006719484.3 XP_006719547.1 11 2898
          XM_006719485.2 XP_006719548.1 10 2340
          XM_006719486.3 XP_006719549.1 10 2328
          XM_006719490.3 XP_006719553.1 13 3802
          XM_011538543.1 XP_011536845.1 13 2665
          XM_011538544.1 XP_011536846.1 13 2653
          XM_011538545.2 XP_011536847.1 12 2633
          XM_011538546.1 XP_011536848.1 14 2736
          XM_011538547.1 XP_011536849.1 14 2780
          XM_011538548.2 XP_011536850.1 11 2426
          XM_011538549.1 XP_011536851.1 14 2747
          XM_011538550.2 XP_011536852.1 13 3736
          XM_011538551.2 XP_011536853.1 13 3724
          XM_011538552.1 XP_011536854.1 13 2739
          XM_011538553.2 XP_011536855.1 13 2669
          XM_017019586.1 XP_016875075.1 12 2621
          XM_017019587.1 XP_016875076.1 11 2554
          XM_017019588.1 XP_016875077.1 14 2724
          XM_017019589.1 XP_016875078.1 13 2708
          XM_017019590.1 XP_016875079.1 14 2768
          XM_017019591.1 XP_016875080.1 12 2959
          XM_017019592.1 XP_016875081.1 11 2413
          XM_017019593.1 XP_016875082.1 14 2788
          XM_017019594.1 XP_016875083.1 13 2650
          XM_017019595.1 XP_016875084.1 13 2754
          XM_017019596.1 XP_016875085.1 12 3743
          XM_017019598.1 XP_016875087.1 12 3731
          XM_017019599.1 XP_016875088.1 13 2703
          XM_017019600.1 XP_016875089.1 12 2976
          XM_017019601.1 XP_016875090.1 13 2727
          XM_017019602.1 XP_016875091.1 14 3795
          XM_017019603.1 XP_016875092.1 12 4059
          XM_017019604.1 XP_016875093.1 10 3411
          XM_017019605.1 XP_016875094.1 10 3399
          GenBank mRNA AF216644.1 - 12 3658
          AF225417.1 - 12 2631
          AF277625.1 - 8 3171
          AK001338.1 - 10 2056
          AK225303.1 - 13 2673
          AL133113.1 - 5 1386
          BC028597.1 - 16 3071
          BC064939.1 - 13 2650 6159768
          BI835735.1 - 1 345

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          single organismal cell-cell adhesion
          myosin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0