Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VEZT SIRNA Assays ›
          Gene Symbol:
          VEZT
          Gene Name:
          vezatin, adherens junctions transmembrane protein
          Gene Aliases:
          VEZATIN
          Chromosome Location:
          Chr.12: 95217746 - 95302799 on Build GRCh38
          UniGene ID:
          Hs.24135
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 24936

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: VEZT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017599.3 NP_060069.3 4 416
          NR_038241.1 5 489
          NR_038242.1 4 412
          XM_006719477.3 XP_006719540.1 4 416
          XM_006719478.2 XP_006719541.1 3 310
          XM_006719479.2 XP_006719542.1 5 487
          XM_006719480.3 XP_006719543.1 5 464
          XM_006719481.2 XP_006719544.1 4 334
          XM_006719482.3 XP_006719545.1 4 715
          XM_006719483.3 XP_006719546.1 4 732
          XM_006719484.3 XP_006719547.1 3 642
          XM_006719490.3 XP_006719553.1 5 487
          XM_011538543.1 XP_011536845.1 5 409
          XM_011538544.1 XP_011536846.1 5 409
          XM_011538545.2 XP_011536847.1 4 377
          XM_011538546.1 XP_011536848.1 6 480
          XM_011538547.1 XP_011536849.1 6 524
          XM_011538549.1 XP_011536851.1 5 409
          XM_011538550.2 XP_011536852.1 5 409
          XM_011538551.2 XP_011536853.1 5 409
          XM_011538552.1 XP_011536854.1 5 483
          XM_011538553.2 XP_011536855.1 5 413
          XM_017019586.1 XP_016875075.1 4 377
          XM_017019587.1 XP_016875076.1 3 310
          XM_017019588.1 XP_016875077.1 6 480
          XM_017019589.1 XP_016875078.1 5 464
          XM_017019590.1 XP_016875079.1 6 524
          XM_017019591.1 XP_016875080.1 4 715
          XM_017019593.1 XP_016875082.1 6 532
          XM_017019594.1 XP_016875083.1 5 394
          XM_017019595.1 XP_016875084.1 4 416
          XM_017019596.1 XP_016875085.1 4 416
          XM_017019597.1 XP_016875086.1 4 416
          XM_017019598.1 XP_016875087.1 4 416
          XM_017019599.1 XP_016875088.1 4 377
          XM_017019600.1 XP_016875089.1 4 732
          XM_017019601.1 XP_016875090.1 5 483
          XM_017019602.1 XP_016875091.1 6 480
          XM_017019603.1 XP_016875092.1 4 732
          GenBank mRNA AF216644.1 - 4 343
          AF225417.1 - 4 387
          AK225303.1 - 5 429
          AK225356.1 - 5 426
          AK307649.1 - 5 483
          AK309943.1 - 4 351
          AL137497.1 - 5 411
          BC028597.1 - 7 737
          BC064939.1 - 5 406 6159768
          DB136255.1 - 4 416

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          single organismal cell-cell adhesion
          myosin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7f76987d59-9ndr4:80/100.66.134.75:80.
          git-commit: fa11bf362e7d47d57dd0debebfa11e830f891645
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0