Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206893.3 | NP_996776.2 | 8 | 2166 | ||
GenBank mRNA | AK122633.1 | - | 8 | 2157 |