Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037442.3 | NP_001032519.1 | 2 | 1015 | ||
NM_001130709.1 | NP_001124181.1 | 2 | 463 | |||
NM_001291993.1 | NP_001278922.1 | 3 | 407 | |||
NM_001291994.1 | NP_001278923.1 | 3 | 1069 | |||
NM_014961.4 | NP_055776.1 | 2 | 1015 | |||
XM_005265656.2 | XP_005265713.1 | 3 | 1053 | |||
XM_005265658.1 | XP_005265715.1 | 2 | 329 | |||
XM_011531750.2 | XP_011530052.1 | 3 | 651 | |||
XM_011531751.2 | XP_011530053.1 | 2 | 664 | |||
XM_011531752.2 | XP_011530054.1 | 2 | 664 | |||
XM_011531753.2 | XP_011530055.1 | 2 | 664 | |||
XM_011531754.2 | XP_011530056.1 | 2 | 664 | |||
XM_011531755.2 | XP_011530057.1 | 2 | 629 | |||
XM_011531756.2 | XP_011530058.1 | 2 | 629 | |||
XM_017007890.1 | XP_016863379.1 | 2 | 664 | |||
XM_017007891.1 | XP_016863380.1 | 3 | 361 | |||
XM_017007892.1 | XP_016863381.1 | 2 | 329 | |||
GenBank mRNA | AB020678.1 | - | 2 | 821 | ||
AF112221.1 | - | 2 | 245 | |||
AK000911.1 | - | 2 | 269 | |||
AK294421.1 | - | 2 | 200 | |||
AK296501.1 | - | 3 | 256 | |||
AK302637.1 | - | 3 | 283 | |||
BC051716.1 | - | 2 | 883 | 5265230 | ||
BC053837.1 | - | 2 | 845 | |||
BC071988.1 | - | 2 | 818 | |||
BC098433.1 | - | 2 | 845 | |||
BM920380.1 | - | 2 | 582 | |||
DA285699.1 | - | 2 | 353 | |||
DB483639.1 | - | 2 | 403 |