Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004273.1 | NP_001004273.1 | 4, 5 | 781 | ||
XM_006247047.2 | XP_006247109.1 | 6, 7 | 989 | |||
XM_006247048.2 | XP_006247110.1 | 6, 7 | 988 | |||
XM_006247049.2 | XP_006247111.1 | 6, 7 | 986 | |||
XM_006247050.2 | XP_006247112.1 | 6, 7 | 985 | |||
XM_006247051.2 | XP_006247113.1 | 5, 6 | 861 | |||
XM_006247052.2 | XP_006247114.1 | 5, 6 | 861 | |||
XM_006247053.2 | XP_006247115.1 | 5, 6 | 858 | |||
XM_006247054.2 | XP_006247116.1 | 5, 6 | 857 | |||
XM_008768070.1 | XP_008766292.1 | 6, 7 | 963 | |||
XM_008768071.1 | XP_008766293.1 | 6, 7 | 933 | |||
GenBank mRNA | AY633742.1 | - | 4, 5 | 781 | ||
FQ220883.1 | - | 2, 3 | 153 | |||
FQ232770.1 | - | 1, 2 | 107 | |||
FQ234201.1 | - | 2, 3 | 248 | |||
FQ234847.1 | - | 1, 2 | 107 |