Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014424.4 | NP_055239.1 | 3 | 1612 | ||
XM_011541249.1 | XP_011539551.1 | 3 | 1633 | |||
XM_011541250.2 | XP_011539552.1 | 3 | 1630 | |||
GenBank mRNA | AF155908.1 | - | 3 | 858 | ||
AJ243191.1 | - | 3 | 858 | |||
AK057295.1 | - | 3 | 1610 | |||
AK075151.1 | - | 2 | 1423 | |||
AK092275.1 | - | 4 | 1135 | |||
AK095911.1 | - | 3 | 865 | |||
AL832181.1 | - | 2 | 1368 | |||
BC006319.2 | - | 3 | 1278 | 4131506 | ||
BX537990.1 | - | 3 | 849 | |||
CR749298.1 | - | 3 | 877 | |||
CR749419.1 | - | 3 | 877 |