Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cysltr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053641.1 | NP_446093.1 | 1 | 928 | ||
XM_006257158.2 | XP_006257220.1 | 3 | 1642 | |||
GenBank mRNA | AB052685.1 | - | 1 | 928 |
Set Membership: |
![]() |