Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TTR SIRNA Assays ›
          Gene Symbol:
          Ttr
          Gene Name:
          transthyretin
          Gene Aliases:
          Lr1, Tbpa
          Chromosome Location:
          Chr.18: 15532963 - 15542180 on Build Rnor_6.0
          UniGene ID:
          Rn.1404
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 200191

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ttr
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012681.2 NP_036813.2 3, 4 455
          XM_006254457.1 XP_006254519.1 4, 5 512
          GenBank mRNA AF479660.1 - 3, 4 390
          BC086946.1 - 3, 4 348 7322736
          FQ209724.1 - 3, 4 373
          FQ209731.1 - 3, 4 373
          FQ209737.1 - 3, 4 373
          FQ209743.1 - 3, 4 373
          FQ209798.1 - 3, 4 373
          FQ209827.1 - 3, 4 372
          FQ209853.1 - 3, 4 373
          FQ209868.1 - 3, 4 373
          FQ209884.1 - 3, 4 373
          FQ209896.1 - 3, 4 366
          FQ209911.1 - 3, 4 373
          FQ209919.1 - 3, 4 372
          FQ209931.1 - 3, 4 370
          FQ209949.1 - 3, 4 373
          FQ209960.1 - 3, 4 372
          FQ209974.1 - 3, 4 373
          FQ209986.1 - 3, 4 373
          FQ209992.1 - 3, 4 370
          FQ210005.1 - 3, 4 373
          FQ210012.1 - 3, 4 372
          FQ210053.1 - 3, 4 373
          FQ210063.1 - 3, 4 373
          FQ210074.1 - 3, 4 368
          FQ210079.1 - 3, 4 373
          FQ210088.1 - 3, 4 373
          FQ210092.1 - 3, 4 373
          FQ210104.1 - 3, 4 372
          FQ210128.1 - 3, 4 373
          FQ210130.1 - 3, 4 373
          FQ210144.1 - 3, 4 373
          FQ210148.1 - 3, 4 373
          FQ210149.1 - 3, 4 373
          FQ210166.1 - 3, 4 373
          FQ210168.1 - 3, 4 373
          FQ210188.1 - 3, 4 373
          FQ210192.1 - 3, 4 368
          FQ210193.1 - 3, 4 373
          FQ210202.1 - 3, 4 373
          FQ210212.1 - 3, 4 373
          FQ210220.1 - 3, 4 373
          FQ210221.1 - 3, 4 373
          FQ210257.1 - 3, 4 373
          FQ210271.1 - 3, 4 373
          FQ210283.1 - 3, 4 370
          FQ210289.1 - 3, 4 368
          FQ210300.1 - 3, 4 373
          FQ210301.1 - 3, 4 373
          FQ210303.1 - 3, 4 373
          FQ210314.1 - 3, 4 373
          FQ210319.1 - 3, 4 373
          FQ210324.1 - 3, 4 371
          FQ210363.1 - 3, 4 373
          FQ210365.1 - 3, 4 372
          FQ210370.1 - 3, 4 373
          FQ210380.1 - 3, 4 373
          FQ210383.1 - 3, 4 370
          FQ210386.1 - 823
          FQ210389.1 - 3, 4 373
          FQ210423.1 - 3, 4 372
          FQ210429.1 - 3, 4 373
          FQ210431.1 - 3, 4 373
          FQ210434.1 - 3, 4 373
          FQ210451.1 - 3, 4 373
          FQ210464.1 - 3, 4 368
          FQ210465.1 - 3, 4 373
          FQ210502.1 - 3, 4 373
          FQ210512.1 - 3, 4 372
          FQ210545.1 - 3, 4 373
          FQ210547.1 - 3, 4 373
          FQ210548.1 - 3, 4 370
          FQ210552.1 - 3, 4 368
          FQ210579.1 - 3, 4 372
          FQ210580.1 - 3, 4 370
          FQ210584.1 - 3, 4 373
          FQ210588.1 - 3, 4 373
          FQ210614.1 - 3, 4 373
          FQ210642.1 - 3, 4 373
          FQ210659.1 - 3, 4 372
          FQ210663.1 - 3, 4 370
          FQ210667.1 - 3, 4 373
          FQ210680.1 - 3, 4 368
          FQ210689.1 - 3, 4 373
          FQ210711.1 - 3, 4 370
          FQ210744.1 - 3, 4 373
          FQ210747.1 - 3, 4 371
          FQ210759.1 - 3, 4 373
          FQ210777.1 - 3, 4 370
          FQ210778.1 - 3, 4 373
          FQ210779.1 - 3, 4 373
          FQ210782.1 - 3, 4 373
          FQ210786.1 - 3, 4 373
          FQ210844.1 - 3, 4 373
          FQ210853.1 - 3, 4 370
          FQ210860.1 - 3, 4 373
          FQ210869.1 - 3, 4 373
          FQ210887.1 - 3, 4 373
          FQ210913.1 - 3, 4 373
          FQ210915.1 - 3, 4 373
          FQ210971.1 - 3, 4 373
          FQ210974.1 - 3, 4 373
          FQ210975.1 - 3, 4 373
          FQ210993.1 - 3, 4 373
          FQ211000.1 - 3, 4 373
          FQ211011.1 - 3, 4 373
          FQ211023.1 - 3, 4 373
          FQ211028.1 - 3, 4 370
          FQ211048.1 - 3, 4 373
          FQ211066.1 - 3, 4 373
          FQ211087.1 - 3, 4 373
          FQ211100.1 - 3, 4 373
          FQ211107.1 - 3, 4 373
          FQ211134.1 - 3, 4 372
          FQ211143.1 - 3, 4 373
          FQ211145.1 - 3, 4 373
          FQ211149.1 - 3, 4 373
          FQ211173.1 - 3, 4 373
          FQ211184.1 - 3, 4 379
          FQ211191.1 - 3, 4 373
          FQ211204.1 - 3, 4 373
          FQ211212.1 - 3, 4 373
          FQ211228.1 - 3, 4 373
          FQ211233.1 - 3, 4 375
          FQ211237.1 - 3, 4 370
          FQ211240.1 - 3, 4 373
          FQ211260.1 - 3, 4 373
          FQ211262.1 - 3, 4 371
          FQ211291.1 - 3, 4 370
          FQ211302.1 - 3, 4 373
          FQ211335.1 - 3, 4 373
          FQ211341.1 - 3, 4 373
          FQ211348.1 - 3, 4 370
          FQ211403.1 - 3, 4 373
          FQ211448.1 - 3, 4 373
          FQ211620.1 - 3, 4 373
          FQ212160.1 - 3, 4 373
          FQ212302.1 - 3, 4 373
          FQ213410.1 - 3, 4 458
          FQ217460.1 - 3, 4 373
          FQ218087.1 - 3, 4 373
          FQ218127.1 - 3, 4 374
          FQ218128.1 - 3, 4 369
          FQ218131.1 - 3, 4 373
          FQ218133.1 - 3, 4 373
          FQ218134.1 - 3, 4 373
          FQ218142.1 - 3, 4 368
          FQ218150.1 - 3, 4 373
          FQ218178.1 - 3, 4 373
          FQ218187.1 - 3, 4 373
          FQ218296.1 - 3, 4 373
          FQ218301.1 - 3, 4 373
          FQ218306.1 - 3, 4 373
          FQ218313.1 - 3, 4 373
          FQ218333.1 - 3, 4 368
          FQ218513.1 - 3, 4 368
          FQ218589.1 - 3, 4 373
          FQ218636.1 - 3, 4 373
          FQ218738.1 - 3, 4 373
          FQ218740.1 - 3, 4 373
          FQ218747.1 - 3, 4 373
          FQ218749.1 - 3, 4 373
          FQ218758.1 - 3, 4 298
          FQ218786.1 - 3, 4 373
          FQ218849.1 - 3, 4 373
          FQ218866.1 - 3, 4 373
          FQ218996.1 - 3, 4 373
          FQ219153.1 - 3, 4 370
          FQ219387.1 - 3, 4 373
          FQ219428.1 - 3, 4 373
          FQ223520.1 - 3, 4 368
          FQ223553.1 - 3, 4 373
          FQ224434.1 - 3, 4 373
          K03252.1 - 3, 4 353
          X14876.1 - 3, 4 346

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          thyroid hormone metabolic process
          retinol metabolic process
          thyroid hormone transport
          hormone activity
          hormone binding
          identical protein binding
          protein heterodimerization activity
          thyroid hormone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0