Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BRCA1 SIRNA Assays ›
          Gene Symbol:
          Brca1
          Gene Name:
          breast cancer 1, early onset
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 89394821 - 89455009 on Build Rnor_6.0
          UniGene ID:
          Rn.217584
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 199803

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Brca1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012514.1 NP_036646.1 4 297
          XM_008768092.1 XP_008766314.1 4 513
          GenBank mRNA AF036760.1 - 4 297
          U60523.1 - 3 161

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          DNA replication
          postreplication repair
          double-strand break repair
          regulation of gene expression by genetic imprinting
          fatty acid biosynthetic process
          cellular response to DNA damage stimulus
          chromosome segregation
          centrosome cycle
          brain development
          response to nutrient
          intrinsic apoptotic signaling pathway in response to DNA damage
          dosage compensation by inactivation of X chromosome
          response to organic substance
          response to ionizing radiation
          positive regulation of vascular endothelial growth factor production
          positive regulation of gene expression
          protein ubiquitination
          positive regulation of protein ubiquitination
          response to estradiol
          response to lipid
          positive regulation of histone acetylation
          negative regulation of histone acetylation
          regulation of cell proliferation
          chordate embryonic development
          response to estrogen
          regulation of DNA methylation
          negative regulation of fatty acid biosynthetic process
          positive regulation of DNA repair
          positive regulation of angiogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of histone H3-K4 methylation
          negative regulation of histone H3-K4 methylation
          negative regulation of histone H3-K9 methylation
          positive regulation of histone H3-K9 methylation
          protein autoubiquitination
          positive regulation of histone H4-K20 methylation
          positive regulation of cell cycle arrest
          cellular response to tumor necrosis factor
          cellular response to indole-3-methanol
          protein K6-linked ubiquitination
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          regulation of cell motility
          negative regulation of reactive oxygen species metabolic process
          positive regulation of histone H3-K9 acetylation
          positive regulation of histone H4-K16 acetylation
          DNA binding
          damaged DNA binding
          RNA binding
          ubiquitin-protein transferase activity
          zinc ion binding
          ligase activity
          enzyme binding
          ubiquitin protein ligase binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-v4tpq:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0