Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133307.1 | NP_579841.1 | 6 | 922 | ||
GenBank mRNA | AF219629.1 | - | 6 | 916 | ||
BC076505.1 | - | 5 | 748 | 7099952 | ||
M18330.1 | - | 6 | 922 |