Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053974.2 | NP_446426.1 | 3 | 179 | ||
XM_008761488.1 | XP_008759710.1 | 4 | 292 | |||
GenBank mRNA | BC087001.1 | - | 3 | 179 | 7122606 | |
FQ218941.1 | - | 3 | 171 | |||
X83399.1 | - | 3 | 214 |