Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021688.3 | NP_067720.1 | 3 | 1920 | ||
XM_006255820.2 | XP_006255882.1 | 3 | 2137 | |||
GenBank mRNA | BC061807.1 | - | 4 | 1600 | 5598415 |