Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009347.2 | NP_033373.2 | 6 | 903 | ||
GenBank mRNA | BC138426.1 | - | 6 | 701 | 8861446 | |
BC171953.1 | - | 6 | 701 | 9054239 | ||
X99805.1 | - | 6 | 903 |