Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019793.3 | NP_062767.3 | 4 | 569 | ||
GenBank mRNA | AB009021.1 | - | 4 | 351 | ||
AB072450.1 | - | 4 | 379 | |||
AF133427.1 | - | 4 | 363 | |||
AF242591.1 | - | 4 | 562 | |||
AK006798.1 | - | 4 | 445 | |||
AK053325.1 | - | 4 | 420 | |||
AK089494.1 | - | 401 | ||||
AK141951.1 | - | 4 | 427 | |||
AK146130.1 | - | 4 | 557 | |||
AK152175.1 | - | 4 | 454 | |||
AK154919.1 | - | 401 | ||||
AK159418.1 | - | 4 | 513 | |||
AK159621.1 | - | 4 | 524 | |||
AK159914.1 | - | 4 | 502 | |||
AK168231.1 | - | 4 | 557 | |||
AK169068.1 | - | 4 | 540 | |||
AK178223.1 | - | 2 | 48 | |||
AK182665.1 | - | 2 | 47 | |||
AK185811.1 | - | 1 | 25 | |||
AK187762.1 | - | 1 | 34 | |||
AK188385.1 | - | 2 | 47 | |||
AK207599.1 | - | 1 | 47 | |||
AK210479.1 | - | 1 | 16 | |||
AK220124.1 | - | 2 | 47 | |||
BC043072.1 | - | 4 | 547 | 6404165 |