Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102407.1 | NP_001095877.1 | 4 | 351 | ||
NM_025875.2 | NP_080151.2 | 4 | 348 | |||
GenBank mRNA | AK009953.1 | - | 4 | 343 | ||
AK010284.1 | - | 4 | 336 | |||
AK082925.1 | - | 4 | 340 | |||
BC020086.1 | - | 4 | 331 | 3671644 | ||
BC058376.1 | - | 4 | 320 | 5709938 | ||
KF051021.1 | - | 4 | 317 |
Gene Symbol: B020018G12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139805.1 | - | 1 | 525 | ||
BC132651.1 | - | 1 | 514 | 40130928 | ||
BC132653.1 | - | 1 | 514 | 40130930 |