Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem50b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030018.3 | NP_084294.1 | 7 | 667 | ||
GenBank mRNA | AK020977.1 | - | 7 | 672 | ||
AK098154.1 | - | 7 | 631 | |||
AK133966.1 | - | 7 | 695 | |||
AK150773.1 | - | 655 | ||||
AK151796.1 | - | 7 | 697 | |||
AK152190.1 | - | 7 | 683 | |||
AK152767.1 | - | 7 | 683 | |||
AK155082.1 | - | 7 | 720 | |||
AK170684.1 | - | 7 | 662 | |||
AK172335.1 | - | 7 | 655 | |||
BC004841.1 | - | 7 | 670 | 2649264 |