Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076676.2 | NP_001070144.1 | 23 | 2697 | ||
NM_001252486.1 | NP_001239415.1 | 23 | 2709 | |||
NM_133247.3 | NP_573510.2 | 23 | 2733 | |||
GenBank mRNA | AF383174.1 | - | 23 | 2722 | ||
AK082251.1 | - | 23 | 2680 | |||
AK082943.1 | - | 23 | 2680 | |||
AK122442.1 | - | 20 | 5191 | |||
AK143844.1 | - | 23 | 2701 | |||
BC005506.1 | - | 23 | 2662 | |||
BC031366.1 | - | 15 | 1849 | |||
BC036980.1 | - | 16 | 1909 | |||
BC049870.1 | - | 10 | 974 | |||
BC067399.1 | - | 21 | 2472 | |||
BC089315.1 | - | 23 | 2655 |