Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147074.2 | NP_667285.2 | 1 | 777 | ||
GenBank mRNA | BC116962.1 | - | 1 | 777 | 40126281 | |
BC120700.1 | - | 1 | 777 | 40129623 |