Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160251.1 | NP_001153723.1 | 2 | 1043 | ||
NM_177643.4 | NP_808311.1 | 2 | 1039 | |||
GenBank mRNA | AK142218.1 | - | 2 | 956 | ||
AK157189.1 | - | 2 | 982 | |||
BC003243.1 | - | 2 | 1000 | 3498439 | ||
BC062153.1 | - | 2 | 993 | 6335572 | ||
CT010200.1 | - | 1 | 874 |