Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198308.1 | NP_938050.1 | 11 | 1582 | ||
GenBank mRNA | AK029575.1 | - | 11 | 1673 | ||
AK078515.1 | - | 5 | 673 | |||
AK129472.1 | - | 11 | 1686 | |||
AY223867.1 | - | 11 | 1582 | |||
BC120745.1 | - | 10 | 1512 | 40129668 | ||
BC125425.1 | - | 10 | 1512 | 40129940 |