Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCSER1 SIRNA Assays ›
Gene Symbol:
CCSER1
Gene Name:
coiled-coil serine rich protein 1
Gene Aliases:
FAM190A
Chromosome Location:
Chr.4: 90127394 - 91602219 on Build GRCh38
UniGene ID:
Hs.654735
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
150009
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCSER1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145065.1
NP_001138537.1
2
508
NM_207491.2
NP_997374.1
2
522
XM_011531936.1
XP_011530238.1
2
647
XM_011531937.2
XP_011530239.1
2
1048
XM_011531938.2
XP_011530240.1
2
372
XM_011531939.2
XP_011530241.1
2
1872
XM_011531941.2
XP_011530243.1
2
647
XM_011531942.2
XP_011530244.1
2
647
XM_011531943.2
XP_011530245.1
2
647
XM_011531945.1
XP_011530247.1
2
647
XM_011531946.2
XP_011530248.1
2
647
XM_011531947.2
XP_011530249.1
2
647
XM_011531948.1
XP_011530250.1
2
647
XM_011531949.2
XP_011530251.1
2
647
XM_011531950.2
XP_011530252.1
2
647
XM_011531951.2
XP_011530253.1
2
647
XM_011531955.2
XP_011530257.1
2
647
XM_011531956.2
XP_011530258.1
2
647
XM_011531957.2
XP_011530259.1
2
647
XM_011531958.2
XP_011530260.1
2
647
XM_017008194.1
XP_016863683.1
2
370
XM_017008195.1
XP_016863684.1
2
647
XM_017008196.1
XP_016863685.1
2
647
XM_017008197.1
XP_016863686.1
2
647
XM_017008198.1
XP_016863687.1
2
647
XM_017008199.1
XP_016863688.1
2
647
XM_017008200.1
XP_016863689.1
2
647
XM_017008201.1
XP_016863690.1
2
647
XM_017008202.1
XP_016863691.1
2
647
GenBank mRNA
AB051467.1
-
2
456
AK097436.1
-
2
508
AK310825.1
-
2
506
BC051694.1
-
2
522
5296943
DB027029.1
-
2
508
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-858577f6d7-cpd7z:80/100.66.134.75:80.
git-commit: 8a4c7f5d60a2609f21a9257632c46d3db2b553cc
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0