Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APBB2 SIRNA Assays ›
          Gene Symbol:
          APBB2
          Gene Name:
          amyloid beta precursor protein binding family B member 2
          Gene Aliases:
          FE65L, FE65L1
          Chromosome Location:
          Chr.4: 40810027 - 41216714 on Build GRCh38
          UniGene ID:
          Hs.479602
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 148182

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APBB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166050.1 NP_001159522.1 18 3041
          NM_001166051.1 NP_001159523.1 8 1062
          NM_001166052.1 NP_001159524.1 7 1056
          NM_001166053.1 NP_001159525.1 8 1058
          NM_001166054.1 NP_001159526.1 7 1052
          NM_004307.1 NP_004298.1 18 3044
          NM_173075.4 NP_775098.2 17 2975
          XM_005248101.3 XP_005248158.1 17 2975
          XM_006714005.3 XP_006714068.1 19 3047
          XM_006714007.3 XP_006714070.1 19 3045
          XM_006714008.3 XP_006714071.1 18 2949
          XM_006714009.3 XP_006714072.1 18 2947
          XM_006714010.3 XP_006714073.1 19 3044
          XM_006714011.3 XP_006714074.1 18 2984
          XM_006714012.3 XP_006714075.1 18 2981
          XM_011513686.2 XP_011511988.1 20 3069
          XM_011513687.2 XP_011511989.1 17 2978
          XM_017008142.1 XP_016863631.1 19 2968
          XM_017008143.1 XP_016863632.1 20 3066
          XM_017008144.1 XP_016863633.1 18 2946
          XM_017008145.1 XP_016863634.1 17 2943
          XM_017008146.1 XP_016863635.1 18 2981
          XM_017008147.1 XP_016863636.1 18 2976
          XM_017008148.1 XP_016863637.1 17 2880
          XM_017008149.1 XP_016863638.1 18 2978
          XM_017008150.1 XP_016863639.1 18 2905
          XM_017008151.1 XP_016863640.1 17 2886
          XM_017008152.1 XP_016863641.1 17 2883
          XM_017008153.1 XP_016863642.1 17 2899
          XM_017008154.1 XP_016863643.1 16 2880
          XM_017008155.1 XP_016863644.1 16 2877
          XM_017008156.1 XP_016863645.1 17 2918
          XM_017008157.1 XP_016863646.1 16 2817
          XM_017008158.1 XP_016863647.1 16 2912
          GenBank mRNA AK226179.1 - 18 2958
          AK289838.1 - 11 1528
          AK299800.1 - 7 1027
          AK304247.1 - 18 2912
          BC027946.1 - 17 2815 5203925
          BC088372.1 - 9 1232

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          regulation of transcription, DNA-templated
          cell cycle arrest
          axon guidance
          extracellular matrix organization
          negative regulation of cell growth
          intracellular signal transduction
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          beta-amyloid binding
          protein binding
          transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0