Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APBB2 SIRNA Assays ›
          Gene Symbol:
          APBB2
          Gene Name:
          amyloid beta precursor protein binding family B member 2
          Gene Aliases:
          FE65L, FE65L1
          Chromosome Location:
          Chr.4: 40810027 - 41216714 on Build GRCh38
          UniGene ID:
          Hs.479602
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 148180

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APBB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166050.1 NP_001159522.1 18 2902
          NM_001166051.1 NP_001159523.1 8 923
          NM_001166052.1 NP_001159524.1 7 917
          NM_001166053.1 NP_001159525.1 8 919
          NM_001166054.1 NP_001159526.1 7 913
          NM_004307.1 NP_004298.1 18 2905
          NM_173075.4 NP_775098.2 17 2836
          XM_005248101.3 XP_005248158.1 17 2836
          XM_006714005.3 XP_006714068.1 19 2908
          XM_006714007.3 XP_006714070.1 19 2906
          XM_006714008.3 XP_006714071.1 18 2810
          XM_006714009.3 XP_006714072.1 18 2808
          XM_006714010.3 XP_006714073.1 19 2905
          XM_006714011.3 XP_006714074.1 18 2845
          XM_006714012.3 XP_006714075.1 18 2842
          XM_011513686.2 XP_011511988.1 20 2930
          XM_011513687.2 XP_011511989.1 17 2839
          XM_017008142.1 XP_016863631.1 19 2829
          XM_017008143.1 XP_016863632.1 20 2927
          XM_017008144.1 XP_016863633.1 18 2807
          XM_017008145.1 XP_016863634.1 17 2804
          XM_017008146.1 XP_016863635.1 18 2842
          XM_017008147.1 XP_016863636.1 18 2837
          XM_017008148.1 XP_016863637.1 17 2741
          XM_017008149.1 XP_016863638.1 18 2839
          XM_017008150.1 XP_016863639.1 18 2766
          XM_017008151.1 XP_016863640.1 17 2747
          XM_017008152.1 XP_016863641.1 17 2744
          XM_017008153.1 XP_016863642.1 17 2760
          XM_017008154.1 XP_016863643.1 16 2741
          XM_017008155.1 XP_016863644.1 16 2738
          XM_017008156.1 XP_016863645.1 17 2779
          XM_017008157.1 XP_016863646.1 16 2678
          XM_017008158.1 XP_016863647.1 16 2773
          GenBank mRNA AK226179.1 - 18 2819
          AK289838.1 - 11 1389
          AK299800.1 - 7 888
          AK304247.1 - 18 2773
          BC027946.1 - 17 2676 5203925
          BC088372.1 - 9 1093
          BX440903.2 - 8 912

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          regulation of transcription, DNA-templated
          cell cycle arrest
          axon guidance
          extracellular matrix organization
          negative regulation of cell growth
          intracellular signal transduction
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          beta-amyloid binding
          protein binding
          transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0