Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCR7 SIRNA Assays ›
          Gene Symbol:
          CCR7
          Gene Name:
          C-C motif chemokine receptor 7
          Gene Aliases:
          BLR2, CC-CKR-7, CCR-7, CD197, CDw197, CMKBR7, EBI1
          Chromosome Location:
          Chr.17: 40553769 - 40565484 on Build GRCh38
          UniGene ID:
          Hs.370036
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146056

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCR7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301714.1 NP_001288643.1 2 119
          NM_001301716.1 NP_001288645.1 3 419
          NM_001301717.1 NP_001288646.1 3 175
          NM_001301718.1 NP_001288647.1 3 265
          NM_001838.3 NP_001829.1 3 169
          GenBank mRNA AK127810.1 - 3 157
          AK310277.1 - 3 419
          AK312732.1 - 3 153
          AY587876.1 - 2 94
          BC035343.1 - 3 135 5180758
          BI910219.1 - 2 94
          DA814913.1 - 3 169
          HY017993.1 - 3 136
          HY021417.1 - 3 266
          L08176.1 - 3 157
          L31581.1 - 3 160

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of T cell polarity
          positive regulation of cell-matrix adhesion
          dendritic cell chemotaxis
          myeloid dendritic cell chemotaxis
          positive regulation of dendritic cell antigen processing and presentation
          positive regulation of hypersensitivity
          positive regulation of humoral immune response
          inflammatory response
          immune response
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          positive regulation of actin filament polymerization
          positive regulation of pseudopodium assembly
          ruffle organization
          response to lipopolysaccharide
          regulation of interferon-gamma production
          positive regulation of interleukin-12 production
          response to prostaglandin E
          chemokine (C-C motif) ligand 19 signaling pathway
          chemokine (C-C motif) ligand 21 signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of phosphatidylinositol 3-kinase activity
          negative thymic T cell selection
          positive regulation of cell adhesion
          positive regulation of protein kinase activity
          positive regulation of JNK cascade
          homeostasis of number of cells
          regulation of interleukin-1 beta secretion
          positive regulation of T cell receptor signaling pathway
          release of sequestered calcium ion into cytosol
          positive regulation of filopodium assembly
          positive regulation of protein kinase B signaling
          positive regulation of ERK1 and ERK2 cascade
          cellular response to cytokine stimulus
          response to nitric oxide
          interleukin-12 secretion
          positive regulation of neutrophil chemotaxis
          activation of GTPase activity
          lymphocyte migration into lymph node
          mature conventional dendritic cell differentiation
          negative regulation of leukocyte apoptotic process
          positive regulation of cell motility
          positive regulation of dendritic cell chemotaxis
          positive regulation of immunological synapse formation
          positive regulation of T cell costimulation
          positive regulation of glycoprotein biosynthetic process involved in immunological synapse formation
          regulation of dendritic cell dendrite assembly
          G-protein coupled receptor activity
          C-C chemokine receptor activity
          chemokine (C-C motif) ligand 19 binding
          chemokine (C-C motif) ligand 21 binding
          C-C motif chemokine 19 receptor activity
          C-C motif chemokine 21 receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0