Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L2 SIRNA Assays ›
Gene Symbol:
EPB41L2
Gene Name:
erythrocyte membrane protein band 4.1 like 2
Gene Aliases:
4.1-G, 4.1G
Chromosome Location:
Chr.6: 130829111 - 131063322 on Build GRCh38
UniGene ID:
Hs.486470
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
145897
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135554.1
NP_001129026.1
2
354
NM_001135555.3
NP_001129027.1
2
367
NM_001199388.2
NP_001186317.1
2
367
NM_001252660.1
NP_001239589.1
2
334
NM_001431.3
NP_001422.1
2
354
XM_006715362.2
XP_006715425.1
2
385
XM_011535524.1
XP_011533826.1
2
354
XM_011535525.1
XP_011533827.1
2
354
XM_011535527.1
XP_011533829.1
2
354
XM_011535528.1
XP_011533830.1
2
354
XM_011535529.1
XP_011533831.1
2
354
XM_011535530.1
XP_011533832.1
2
354
XM_011535534.1
XP_011533836.1
2
354
XM_011535536.1
XP_011533838.1
2
354
XM_017010349.1
XP_016865838.1
2
382
XM_017010350.1
XP_016865839.1
2
316
XM_017010351.1
XP_016865840.1
2
378
XM_017010352.1
XP_016865841.1
2
220
XM_017010353.1
XP_016865842.1
2
354
XM_017010354.1
XP_016865843.1
2
354
XM_017010355.1
XP_016865844.1
2
382
XM_017010356.1
XP_016865845.1
2
354
XM_017010357.1
XP_016865846.1
2
316
XM_017010358.1
XP_016865847.1
2
354
XM_017010359.1
XP_016865848.1
2
382
XM_017010360.1
XP_016865849.1
2
220
XM_017010361.1
XP_016865850.1
2
382
XM_017010362.1
XP_016865851.1
2
316
XM_017010363.1
XP_016865852.1
2
344
GenBank mRNA
AB209522.1
-
2
265
AF027299.1
-
2
216
AK295124.1
-
2
296
AK307764.1
-
2
387
AL698619.1
-
2
278
AY047584.1
-
1
180
BI461477.1
-
2
328
CR749262.1
-
2
296
CR936793.1
-
2
334
DA108957.1
-
2
354
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cortical actin cytoskeleton organization
actin binding
structural molecule activity
spectrin binding
PH domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c48b94556-6l4lq:80/100.66.134.75:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0