Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VEZT SIRNA Assays ›
Gene Symbol:
VEZT
Gene Name:
vezatin, adherens junctions transmembrane protein
Gene Aliases:
VEZATIN
Chromosome Location:
Chr.12: 95217746 - 95302799 on Build GRCh38
UniGene ID:
Hs.24135
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140405
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VEZT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017599.3
NP_060069.3
12
1983
NR_038241.1
13
2056
NR_038242.1
12
1991
XM_006719477.3
XP_006719540.1
12
1995
XM_006719478.2
XP_006719541.1
11
1889
XM_006719479.2
XP_006719542.1
13
2066
XM_006719480.3
XP_006719543.1
13
2043
XM_006719481.2
XP_006719544.1
12
1913
XM_006719482.3
XP_006719545.1
12
2294
XM_006719483.3
XP_006719546.1
12
2311
XM_006719484.3
XP_006719547.1
11
2221
XM_006719485.2
XP_006719548.1
10
1663
XM_006719486.3
XP_006719549.1
10
1651
XM_006719490.3
XP_006719553.1
13
3125
XM_011538543.1
XP_011536845.1
13
1988
XM_011538544.1
XP_011536846.1
13
1976
XM_011538545.2
XP_011536847.1
12
1956
XM_011538546.1
XP_011536848.1
14
2059
XM_011538547.1
XP_011536849.1
14
2103
XM_011538548.2
XP_011536850.1
11
1749
XM_011538549.1
XP_011536851.1
14
2070
XM_011538550.2
XP_011536852.1
13
3059
XM_011538551.2
XP_011536853.1
13
3047
XM_011538552.1
XP_011536854.1
13
2062
XM_011538553.2
XP_011536855.1
13
1992
XM_017019586.1
XP_016875075.1
12
1944
XM_017019587.1
XP_016875076.1
11
1877
XM_017019588.1
XP_016875077.1
14
2047
XM_017019589.1
XP_016875078.1
13
2031
XM_017019590.1
XP_016875079.1
14
2091
XM_017019591.1
XP_016875080.1
12
2282
XM_017019592.1
XP_016875081.1
11
1736
XM_017019593.1
XP_016875082.1
14
2111
XM_017019594.1
XP_016875083.1
13
1973
XM_017019595.1
XP_016875084.1
13
2077
XM_017019596.1
XP_016875085.1
12
3066
XM_017019597.1
XP_016875086.1
13
2065
XM_017019598.1
XP_016875087.1
12
3054
XM_017019599.1
XP_016875088.1
13
2026
XM_017019600.1
XP_016875089.1
12
2299
XM_017019601.1
XP_016875090.1
13
2050
XM_017019602.1
XP_016875091.1
14
3118
XM_017019603.1
XP_016875092.1
12
3382
XM_017019604.1
XP_016875093.1
10
2734
XM_017019605.1
XP_016875094.1
10
2722
GenBank mRNA
AF216644.1
-
12
2982
AF225417.1
-
12
1954
AF277625.1
-
8
2494
AK001338.1
-
10
1379
AK225303.1
-
13
1996
AK225356.1
-
13
2005
AK307649.1
-
13
2062
AL133113.1
-
5
709
BC028597.1
-
16
2394
BC064939.1
-
13
1973
6159768
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
single organismal cell-cell adhesion
myosin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7f76987d59-9ndr4:80/100.66.134.75:80.
git-commit: fa11bf362e7d47d57dd0debebfa11e830f891645
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0