Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NISCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007184.3 | NP_009115.2 | 13, 14 | 1670 | ||
XM_005264839.2 | XP_005264896.1 | 12, 13 | 1519 | |||
XM_017005643.1 | XP_016861132.1 | 4, 5 | 511 | |||
GenBank mRNA | AB023192.1 | - | 14, 15 | 1608 | ||
AF058290.1 | - | 1, 2 | 133 | |||
AF082516.1 | - | 13, 14 | 1562 | |||
AK027428.1 | - | 13, 14 | 1548 | |||
AK291398.1 | - | 13, 14 | 1574 | |||
AK302441.1 | - | 4, 5 | 558 | |||
AK315001.1 | - | 13, 14 | 1574 | |||
BC038102.2 | - | 13, 14 | 1574 | 5268247 | ||
BC054494.1 | - | 13, 14 | 1568 | 6195532 | ||
BC056900.1 | - | 13, 14 | 1540 | 6143316 |