Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NOL4 SIRNA Assays ›
          Gene Symbol:
          NOL4
          Gene Name:
          nucleolar protein 4
          Gene Aliases:
          CT125, HRIHFB2255, NOLP
          Chromosome Location:
          Chr.18: 33851100 - 34224834 on Build GRCh38
          UniGene ID:
          Hs.514795
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 13572

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NOL4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198546.1 NP_001185475.1 4, 5 945
          NM_001198547.1 NP_001185476.1 4, 5 834
          NM_001198548.1 NP_001185477.1 4, 5 945
          NM_001282527.1 NP_001269456.1 5, 6 875
          NM_003787.4 NP_003778.2 4, 5 945
          XM_006722563.2 XP_006722626.1 5, 6 1702
          XM_006722564.2 XP_006722627.1 4, 5 687
          XM_006722565.1 XP_006722628.1 5, 6 721
          XM_011526237.1 XP_011524539.1 5, 6 1699
          XM_011526238.1 XP_011524540.1 4, 5 1311
          XM_011526239.2 XP_011524541.1 3, 4 703
          XM_017026052.1 XP_016881541.1 3, 4 533
          XM_017026053.1 XP_016881542.1 4, 5 685
          XM_017026054.1 XP_016881543.1 5, 6 813
          XM_017026055.1 XP_016881544.1 5, 6 1352
          XM_017026056.1 XP_016881545.1 5, 6 606
          XM_017026057.1 XP_016881546.1 4, 5 1065
          XM_017026058.1 XP_016881547.1 4, 5 424
          XM_017026059.1 XP_016881548.1 4, 5 2227
          GenBank mRNA AB017800.1 - 4, 5 955
          AK091463.1 - 5, 6 724
          AK297185.1 - 4, 5 599
          AK299850.1 - 4, 5 456
          AK312562.1 - 5, 6 472
          BC000313.2 - 4, 5 631 2821116
          BT006763.1 - 3, 4 305
          CR456730.1 - 3, 4 305

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0