Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZSCAN5A SIRNA Assays ›
Gene Symbol:
ZSCAN5A
Gene Name:
zinc finger and SCAN domain containing 5A
Gene Aliases:
ZNF495, ZSCAN5
Chromosome Location:
Chr.19: 56221303 - 56368383 on Build GRCh38
UniGene ID:
Hs.177688
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
130563
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZSCAN5A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001322061.1
NP_001308990.1
7
1727
NM_001322062.1
NP_001308991.1
6
1495
NM_001322064.1
NP_001308993.1
6
1667
NM_001322065.1
NP_001308994.1
6
1542
NM_001322066.1
NP_001308995.1
6
1737
NM_001322067.1
NP_001308996.1
6
1441
NM_001322068.1
NP_001308997.1
6
1566
NM_001322069.1
NP_001308998.1
6
1737
NM_001322070.1
NP_001308999.1
6
1612
NM_001322072.1
NP_001309001.1
7
1889
NM_001322073.1
NP_001309002.1
6
1539
NM_001322074.1
NP_001309003.1
6
1666
NM_001322075.1
NP_001309004.1
6
1734
NM_001322076.1
NP_001309005.1
6
1609
NM_001322077.1
NP_001309006.1
5
1156
NM_001322078.1
NP_001309007.1
5
1117
NM_024303.2
NP_077279.1
6
1623
XM_005259254.1
XP_005259311.1
6
1498
XM_006723377.3
XP_006723440.1
5
1779
XM_011527308.1
XP_011525610.1
5
1307
XM_017027298.1
XP_016882787.1
5
1767
XM_017027299.1
XP_016882788.1
5
1861
GenBank mRNA
AK098700.1
-
5
1252
AK223485.1
-
4
1320
AK301873.1
-
5
1156
BC002636.1
-
5
1359
3606994
BC043232.1
-
7
1894
5295665
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0