Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WDR27 SIRNA Assays ›
Gene Symbol:
WDR27
Gene Name:
WD repeat domain 27
Gene Aliases:
-
Chromosome Location:
Chr.6: 169426420 - 169702071 on Build GRCh38
UniGene ID:
Hs.131903
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
128500
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WDR27
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001202550.1
NP_001189479.1
2, 3
717
NM_182552.4
NP_872358.4
2, 3
717
XM_011535682.2
XP_011533984.1
3, 4
907
XM_011535684.2
XP_011533986.1
3, 4
907
XM_011535685.2
XP_011533987.1
3, 4
907
XM_011535687.2
XP_011533989.1
3, 4
907
XM_011535688.2
XP_011533990.1
3, 4
907
XM_011535689.2
XP_011533991.1
3, 4
907
XM_011535690.2
XP_011533992.1
3, 4
907
XM_011535691.2
XP_011533993.1
3, 4
907
XM_011535692.2
XP_011533994.1
3, 4
907
XM_011535693.2
XP_011533995.1
3, 4
907
XM_011535694.2
XP_011533996.1
3, 4
907
XM_011535696.2
XP_011533998.1
3, 4
907
XM_011535697.2
XP_011533999.1
3, 4
907
XM_017010658.1
XP_016866147.1
3, 4
907
XM_017010659.1
XP_016866148.1
3, 4
907
XM_017010660.1
XP_016866149.1
3, 4
907
XM_017010661.1
XP_016866150.1
3, 4
907
XM_017010662.1
XP_016866151.1
3, 4
907
XM_017010663.1
XP_016866152.1
3, 4
907
XM_017010664.1
XP_016866153.1
3, 4
907
XM_017010665.1
XP_016866154.1
3, 4
907
XM_017010666.1
XP_016866155.1
3, 4
907
XM_017010667.1
XP_016866156.1
3, 4
907
XM_017010668.1
XP_016866157.1
3, 4
907
XM_017010669.1
XP_016866158.1
2, 3
725
XM_017010670.1
XP_016866159.1
3, 4
907
XM_017010671.1
XP_016866160.1
3, 4
907
XM_017010672.1
XP_016866161.1
3, 4
907
XM_017010673.1
XP_016866162.1
3, 4
907
XM_017010674.1
XP_016866163.1
3, 4
907
XM_017010675.1
XP_016866164.1
2, 3
725
GenBank mRNA
AK293148.1
-
2, 3
683
BC131633.1
-
2, 3
240
40119040
BC142982.1
-
2, 3
307
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0