Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM161B SIRNA Assays ›
Gene Symbol:
TMEM161B
Gene Name:
transmembrane protein 161B
Gene Aliases:
FLB3342, PRO1313
Chromosome Location:
Chr.5: 88189419 - 88268917 on Build GRCh38
UniGene ID:
Hs.379972
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
128185
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM161B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289007.1
NP_001275936.1
10
1089
NM_001289008.1
NP_001275937.1
11
1221
NM_153354.4
NP_699185.1
10
1089
NR_110266.1
11
1189
XM_006714553.1
XP_006714616.1
10
1039
XM_006714554.3
XP_006714617.1
9
976
XM_006714555.3
XP_006714618.1
8
858
XM_006714556.3
XP_006714619.2
8
819
XM_011543201.2
XP_011541503.1
9
956
XM_011543202.2
XP_011541504.2
8
1501
XM_011543203.2
XP_011541505.2
6
1306
XM_011543204.2
XP_011541506.2
8
774
XM_017009093.1
XP_016864582.1
10
1039
XM_017009094.1
XP_016864583.1
9
930
XM_017009095.1
XP_016864584.1
10
1039
XM_017009096.1
XP_016864585.1
9
960
XM_017009097.1
XP_016864586.1
9
963
XM_017009098.1
XP_016864587.1
9
908
XM_017009099.1
XP_016864588.1
8
837
XM_017009100.1
XP_016864589.1
9
930
XM_017009101.1
XP_016864590.1
9
892
XM_017009102.1
XP_016864591.1
9
893
XM_017009103.1
XP_016864592.1
9
960
XM_017009104.1
XP_016864593.1
9
1023
XM_017009105.1
XP_016864594.1
10
1001
XM_017009106.1
XP_016864595.1
9
893
XM_017009107.1
XP_016864596.1
9
923
XM_017009108.1
XP_016864597.1
8
808
XM_017009109.1
XP_016864598.1
9
973
XM_017009110.1
XP_016864599.1
9
959
GenBank mRNA
AK300893.1
-
11
1171
AK301091.1
-
10
1040
BC037287.2
-
10
1058
4837154
BM479365.1
-
7
698
BX647345.1
-
11
1109
BX648757.1
-
8
4862
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0