Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1324L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142749.2 | NP_001136221.1 | 7 | 1075 | ||
NM_001291990.1 | NP_001278919.1 | 7 | 833 | |||
NM_001291991.1 | NP_001278920.1 | 6 | 550 | |||
NM_152748.3 | NP_689961.3 | 6 | 672 | |||
XM_006715894.3 | XP_006715957.1 | 7 | 775 | |||
XM_011515918.2 | XP_011514220.1 | 7 | 926 | |||
XM_011515921.1 | XP_011514223.1 | 6 | 948 | |||
XM_017011843.1 | XP_016867332.1 | 7 | 1071 | |||
XM_017011844.1 | XP_016867333.1 | 6 | 665 | |||
GenBank mRNA | AK055902.1 | - | 6 | 531 | ||
AK296250.1 | - | 6 | 589 | |||
AK303619.1 | - | 7 | 835 | |||
BC117309.1 | - | 3 | 220 | 40125860 |