Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270384.1 | NP_001257313.1 | 3 | 373 | ||
NM_198549.3 | NP_940951.1 | 3 | 373 | |||
XM_005270846.4 | XP_005270903.1 | 3 | 367 | |||
XM_005270847.4 | XP_005270904.1 | 3 | 1051 | |||
XM_005270848.4 | XP_005270905.1 | 3 | 660 | |||
XM_011541422.2 | XP_011539724.1 | 3 | 424 | |||
XM_017001227.1 | XP_016856716.1 | 3 | 343 | |||
XM_017001228.1 | XP_016856717.1 | 3 | 660 | |||
XM_017001229.1 | XP_016856718.1 | 3 | 435 | |||
XM_017001230.1 | XP_016856719.1 | 3 | 448 | |||
XM_017001231.1 | XP_016856720.1 | 3 | 367 | |||
XM_017001232.1 | XP_016856721.1 | 3 | 425 | |||
GenBank mRNA | AK092412.1 | - | 3 | 341 | ||
AK296411.1 | - | 2 | 234 | |||
BC130387.1 | - | 3 | 361 | 40146418 | ||
BC144167.1 | - | 3 | 361 | 9052687 | ||
BX537792.1 | - | 3 | 373 |