Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139459.1 | NP_001132931.1 | 3 | 940 | ||
NM_152609.2 | NP_689822.2 | 3 | 814 | |||
XM_005273081.3 | XP_005273138.2 | 3 | 599 | |||
XM_011544110.2 | XP_011542412.1 | 3 | 665 | |||
XM_011544111.1 | XP_011542413.1 | 3 | 836 | |||
XM_011544112.1 | XP_011542414.1 | 2 | 383 | |||
XM_011544114.2 | XP_011542416.1 | 2 | 273 | |||
XM_017000487.1 | XP_016855976.1 | 4 | 854 | |||
XM_017000488.1 | XP_016855977.1 | 3 | 647 | |||
XM_017000489.1 | XP_016855978.1 | 3 | 714 | |||
GenBank mRNA | AK056563.1 | - | 3 | 647 | ||
AK074142.1 | - | 3 | 577 | |||
AL834246.2 | - | 3 | 648 | |||
BC010228.2 | - | 3 | 816 | 4151228 | ||
BC036200.1 | - | 3 | 707 | |||
BX647629.1 | - | 3 | 599 |