Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167606.1 | NP_001161078.1 | 4 | 497 | ||
NM_006861.6 | NP_006852.1 | 4 | 497 | |||
XM_011537779.1 | XP_011536081.1 | 4 | 434 | |||
XM_017018723.1 | XP_016874212.1 | 4 | 399 | |||
GenBank mRNA | AF498960.1 | - | 4 | 308 | ||
AK125543.1 | - | 5 | 1272 | |||
AK298391.1 | - | 1 | 17 | |||
AK304620.1 | - | 4 | 404 | |||
AK312538.1 | - | 4 | 407 | |||
BC015931.2 | - | 4 | 412 | 3907209 | ||
BM467132.1 | - | 4 | 479 | |||
BQ233709.1 | - | 4 | 453 | |||
BT020024.1 | - | 4 | 308 | |||
CR536486.1 | - | 4 | 308 | |||
CR541683.1 | - | 4 | 308 | |||
DC349830.1 | - | 4 | 494 | |||
DC370610.1 | - | 4 | 497 | |||
DC408044.1 | - | 4 | 497 | |||
X79781.1 | - | 4 | 389 |