Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF415 SIRNA Assays ›
Gene Symbol:
ZNF415
Gene Name:
zinc finger protein 415
Gene Aliases:
ZfLp
Chromosome Location:
Chr.19: 53107879 - 53133077 on Build GRCh38
UniGene ID:
Hs.147765
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116214
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF415
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136038.2
NP_001129510.2
4
2162
NM_001164309.1
NP_001157781.1
4
2035
NM_018355.3
NP_060825.2
4
1930
NR_028343.1
7
2291
XM_006723267.1
XP_006723330.1
6
2248
XM_006723268.1
XP_006723331.1
4
1996
XM_006723272.2
XP_006723335.1
5
2028
XM_006723273.2
XP_006723336.1
5
2112
XM_006723274.2
XP_006723337.1
5
2189
XM_006723277.2
XP_006723340.1
5
2123
XM_006723279.1
XP_006723342.1
5
2282
XM_006723280.2
XP_006723343.1
2
1738
XM_006723281.2
XP_006723344.1
3
1764
XM_006723282.1
XP_006723345.1
3
2073
XM_011527100.1
XP_011525402.1
5
2272
XM_011527101.2
XP_011525403.1
5
2247
XM_011527103.1
XP_011525405.1
5
2067
XM_011527104.1
XP_011525406.1
5
2361
XM_017026952.1
XP_016882441.1
5
2075
XM_017026953.1
XP_016882442.1
6
2395
XM_017026954.1
XP_016882443.1
6
2272
XM_017026955.1
XP_016882444.1
6
2223
XM_017026956.1
XP_016882445.1
7
2373
XM_017026957.1
XP_016882446.1
6
2111
XM_017026958.1
XP_016882447.1
7
2427
XM_017026959.1
XP_016882448.1
6
2298
XM_017026960.1
XP_016882449.1
5
2013
XM_017026961.1
XP_016882450.1
6
2459
XM_017026962.1
XP_016882451.1
6
2304
XM_017026963.1
XP_016882452.1
5
2200
XM_017026964.1
XP_016882453.1
5
2150
XM_017026965.1
XP_016882454.1
4
2164
XM_017026966.1
XP_016882455.1
5
2292
XM_017026967.1
XP_016882456.1
4
2090
XM_017026968.1
XP_016882457.1
6
2388
XM_017026969.1
XP_016882458.1
2
1809
XM_017026970.1
XP_016882459.1
6
2265
XM_017026971.1
XP_016882460.1
6
2484
XM_017026972.1
XP_016882461.1
4
2066
XM_017026973.1
XP_016882462.1
5
2258
XM_017026974.1
XP_016882463.1
6
2337
GenBank mRNA
AF038179.1
-
1
738
AK002053.1
-
4
1902
AK095504.1
-
7
2263
AY283600.1
-
4
1902
BC063880.1
-
5
2113
30388597
BC069668.1
-
5
2112
7262632
DQ925695.1
-
3
1764
DQ925696.1
-
5
2101
DQ925697.1
-
6
2111
DQ925698.1
-
5
2213
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c48b94556-6l4lq:80/100.66.134.75:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0