Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF415 SIRNA Assays ›
          Gene Symbol:
          ZNF415
          Gene Name:
          zinc finger protein 415
          Gene Aliases:
          ZfLp
          Chromosome Location:
          Chr.19: 53107879 - 53133077 on Build GRCh38
          UniGene ID:
          Hs.147765
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 116213

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF415
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136038.2 NP_001129510.2 4 741
          NM_001164309.1 NP_001157781.1 4 614
          NM_018355.3 NP_060825.2 4 509
          NR_028343.1 7 870
          XM_006723267.1 XP_006723330.1 6 827
          XM_006723268.1 XP_006723331.1 4 575
          XM_006723272.2 XP_006723335.1 5 607
          XM_006723273.2 XP_006723336.1 5 691
          XM_006723274.2 XP_006723337.1 5 768
          XM_006723277.2 XP_006723340.1 5 702
          XM_006723279.1 XP_006723342.1 5 861
          XM_006723280.2 XP_006723343.1 2 317
          XM_006723281.2 XP_006723344.1 3 343
          XM_006723282.1 XP_006723345.1 3 652
          XM_011527100.1 XP_011525402.1 5 851
          XM_011527101.2 XP_011525403.1 5 826
          XM_011527103.1 XP_011525405.1 5 646
          XM_011527104.1 XP_011525406.1 5 940
          XM_017026952.1 XP_016882441.1 5 654
          XM_017026953.1 XP_016882442.1 6 974
          XM_017026954.1 XP_016882443.1 6 851
          XM_017026955.1 XP_016882444.1 6 802
          XM_017026956.1 XP_016882445.1 7 952
          XM_017026957.1 XP_016882446.1 6 690
          XM_017026958.1 XP_016882447.1 7 1006
          XM_017026959.1 XP_016882448.1 6 877
          XM_017026960.1 XP_016882449.1 5 592
          XM_017026961.1 XP_016882450.1 6 1038
          XM_017026962.1 XP_016882451.1 6 883
          XM_017026963.1 XP_016882452.1 5 779
          XM_017026964.1 XP_016882453.1 5 729
          XM_017026965.1 XP_016882454.1 4 743
          XM_017026966.1 XP_016882455.1 5 871
          XM_017026967.1 XP_016882456.1 4 669
          XM_017026968.1 XP_016882457.1 6 967
          XM_017026969.1 XP_016882458.1 2 388
          XM_017026970.1 XP_016882459.1 6 844
          XM_017026971.1 XP_016882460.1 6 1063
          XM_017026972.1 XP_016882461.1 4 645
          XM_017026973.1 XP_016882462.1 5 837
          XM_017026974.1 XP_016882463.1 6 916
          GenBank mRNA AK002053.1 - 4 481
          AK095504.1 - 7 842
          AL534796.3 - 5 691
          AY283600.1 - 4 481
          BC063880.1 - 5 692 30388597
          BC069668.1 - 5 691 7262632
          DA367299.1 - 3 481
          DQ925695.1 - 3 343
          DQ925696.1 - 5 680
          DQ925697.1 - 6 690
          DQ925698.1 - 5 792

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c48b94556-6l4lq:80/100.66.134.75:80.
          git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0